Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005934.3 | NP_005925.2 | 11, 12 | 1718 | ||
XM_011528021.1 | XP_011526323.1 | 11, 12 | 1692 | |||
XM_011528022.1 | XP_011526324.1 | 11, 12 | 1575 | |||
XM_011528023.1 | XP_011526325.1 | 10, 11 | 1566 | |||
XM_017026819.1 | XP_016882308.1 | 11, 12 | 2274 | |||
GenBank mRNA | D14539.1 | - | 11, 12 | 1718 | ||
L04285.1 | - | 10, 11 | 1554 |