Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810026J23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178619.4 | NP_848734.1 | 2 | 818 | ||
GenBank mRNA | AK028799.1 | - | 2 | 649 | ||
AK030602.1 | - | 2 | 671 | |||
AK034762.1 | - | 2 | 515 | |||
AK088045.1 | - | 2 | 591 | |||
AK089121.1 | - | 2 | 577 | |||
BC025883.1 | - | 2 | 576 | |||
BC057071.1 | - | 2 | 818 | 5695590 |