Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AFDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040000.2 | NP_001035089.1 | 29 | 4479 | ||
NM_001207008.1 | NP_001193937.1 | 28 | 4428 | |||
NM_001291964.1 | NP_001278893.1 | 29 | 4533 | |||
XM_005266996.3 | XP_005267053.1 | 29 | 4598 | |||
XM_005266997.2 | XP_005267054.1 | 29 | 4596 | |||
XM_006715491.2 | XP_006715554.1 | 30 | 4617 | |||
XM_006715494.3 | XP_006715557.1 | 28 | 4533 | |||
XM_011535842.2 | XP_011534144.1 | 29 | 4598 | |||
XM_017010878.1 | XP_016866367.1 | 30 | 4617 | |||
XM_017010879.1 | XP_016866368.1 | 30 | 4619 | |||
XM_017010880.1 | XP_016866369.1 | 30 | 4598 | |||
XM_017010881.1 | XP_016866370.1 | 30 | 4599 | |||
XM_017010882.1 | XP_016866371.1 | 29 | 4596 | |||
XM_017010883.1 | XP_016866372.1 | 29 | 4572 | |||
XM_017010884.1 | XP_016866373.1 | 29 | 4574 | |||
XM_017010885.1 | XP_016866374.1 | 28 | 4533 | |||
XM_017010886.1 | XP_016866375.1 | 30 | 4617 | |||
XM_017010887.1 | XP_016866376.1 | 27 | 3856 | |||
GenBank mRNA | AB209420.1 | - | 29 | 4302 | ||
AB621809.1 | - | 29 | 4533 | |||
AL161973.1 | - | 22 | 3335 | |||
BX649158.1 | - | 14 | 2927 | |||
U02478.1 | - | 28 | 4289 |