Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027275.3 | NP_081551.2 | 17, 18 | 1395 | ||
GenBank mRNA | AK013131.1 | - | 3, 4 | 174 | ||
AK146005.1 | - | 17, 18 | 1384 | |||
AK172480.1 | - | 18, 19 | 1460 | |||
BC026759.1 | - | 1, 2 | 50 | |||
BC055844.1 | - | 3, 4 | 222 | |||
BC060687.1 | - | 8, 9 | 616 | |||
BC084595.1 | - | 8, 9 | 645 | |||
BC115434.2 | - | 17, 18 | 1391 | 40094203 |