Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252020.1 | NP_001238949.1 | 25 | 3719 | ||
NM_001252024.1 | NP_001238953.1 | 26 | 3507 | |||
NM_002420.5 | NP_002411.3 | 25 | 3421 | |||
GenBank mRNA | AB115498.1 | - | 22 | 3078 | ||
AB115499.1 | - | 22 | 3078 | |||
AB115500.1 | - | 21 | 2933 | |||
AB115501.1 | - | 22 | 3078 | |||
AB115502.1 | - | 22 | 3060 | |||
AF071787.1 | - | 25 | 3398 | |||
HM135790.1 | - | 26 | 3502 | |||
HM135791.1 | - | 25 | 3719 |