Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291002.1 | NP_001277931.1 | 4 | 418 | ||
NM_175154.3 | NP_780363.1 | 4 | 524 | |||
GenBank mRNA | AK053002.1 | - | 4 | 374 | ||
AK082521.1 | - | 4 | 415 | |||
AK152851.1 | - | 2 | 108 | |||
AK154762.1 | - | 4 | 373 | |||
AK171939.1 | - | 4 | 401 | |||
AK176923.1 | - | 4 | 301 | |||
AK179543.1 | - | 2 | 181 | |||
AK187716.1 | - | 1 | 24 | |||
BC079843.1 | - | 4 | 388 | 5690056 |