Gene Symbol: TRPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252020.1 | NP_001238949.1 | 12 | 1900 | ||
NM_001252024.1 | NP_001238953.1 | 13 | 1688 | |||
NM_002420.5 | NP_002411.3 | 12 | 1602 | |||
GenBank mRNA | AB115498.1 | - | 9 | 1259 | ||
AB115499.1 | - | 9 | 1259 | |||
AB115500.1 | - | 9 | 1289 | |||
AB115501.1 | - | 9 | 1259 | |||
AB115502.1 | - | 9 | 1241 | |||
AF071787.1 | - | 12 | 1579 | |||
HM135790.1 | - | 13 | 1683 | |||
HM135791.1 | - | 12 | 1900 |