Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142950.1 | NP_001136422.1 | 14 | 1629 | ||
NM_027350.3 | NP_081626.2 | 14 | 1626 | |||
GenBank mRNA | AK013880.1 | - | 5 | 606 | ||
AK077699.1 | - | 14 | 1607 | |||
AK157244.1 | - | 14 | 1589 | |||
AK172657.1 | - | 14 | 1588 | |||
BC005473.1 | - | 13 | 1841 | |||
BC052849.1 | - | 14 | 1587 | 30057822 |