Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Secisbp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177608.3 | NP_808276.2 | 5 | 927 | ||
GenBank mRNA | AK049924.1 | - | 5 | 928 | ||
AK145135.1 | - | 5 | 813 | |||
AK169713.1 | - | 5 | 814 | |||
AK172920.1 | - | 5 | 796 | |||
BC095939.1 | - | 5 | 794 | |||
BC141056.1 | - | 5 | 887 | 9055668 |