Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190885.1 | NP_001177814.1 | 4 | 630 | ||
NM_001190886.1 | NP_001177815.1 | 3 | 586 | |||
NM_001290690.1 | NP_001277619.1 | 3 | 785 | |||
NM_027398.3 | NP_081674.2 | 3 | 597 | |||
GenBank mRNA | AB075041.1 | - | 3 | 227 | ||
AJ278534.1 | - | 2 | 155 | |||
AK014313.1 | - | 3 | 586 | |||
AK045948.1 | - | 4 | 631 | |||
AK046398.1 | - | 4 | 619 | |||
AK048178.1 | - | 2 | 1478 | |||
AK081845.1 | - | 3 | 587 | |||
AK082267.1 | - | 2 | 1316 | |||
AY050525.1 | - | 4 | 235 | |||
AY050526.1 | - | 3 | 202 | |||
AY171234.1 | - | 3 | 661 | |||
AY647242.1 | - | 3 | 429 | |||
BC034241.1 | - | 3 | 539 | 4503251 | ||
BU702946.1 | - | 2 | 283 | |||
DQ148493.1 | - | 3 | 539 | |||
DQ148494.1 | - | 3 | 587 | |||
DQ148495.1 | - | 4 | 235 |