Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ica1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027407.3 | NP_081683.3 | 2 | 368 | ||
GenBank mRNA | AK029991.1 | - | 2 | 140 | ||
AK158859.1 | - | 2 | 204 | |||
AK165131.1 | - | 2 | 194 | |||
BC079669.1 | - | 2 | 368 | 30355976 |