Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011942.1 | - | 4 | 1041 | ||
AK053363.1 | - | 5 | 1143 | |||
AK150539.1 | - | 5 | 1302 | |||
AK151163.1 | - | 5 | 1302 | |||
AK154611.1 | - | 4 | 1110 | |||
BC020064.1 | - | 5 | 1208 | |||
BC048708.1 | - | 4 | 933 | |||
BC060191.1 | - | 5 | 1177 | |||
BC063765.1 | - | 4 | 976 | |||
BC080823.1 | - | 4 | 986 | |||
BC116944.1 | - | 4 | 1084 | 40126263 | ||
BC116970.1 | - | 4 | 1084 | 40126289 | ||
BC144746.1 | - | 4 | 985 |