Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291134.1 | NP_001278063.1 | 2 | 574 | ||
NM_001291135.1 | NP_001278064.1 | 2 | 574 | |||
NM_027434.3 | NP_081710.1 | 2 | 574 | |||
GenBank mRNA | AK011979.1 | - | 2 | 575 | ||
AK035806.1 | - | 2 | 564 | |||
AK139995.1 | - | 273 | ||||
AK147718.1 | - | 2 | 547 | |||
AK147930.1 | - | 2 | 536 | |||
AK152125.1 | - | 2 | 372 | |||
AK152220.1 | - | 2 | 372 | |||
BC028819.1 | - | 2 | 356 | 5100766 | ||
BC042447.1 | - | 2 | 286 | 5357316 | ||
BC054350.1 | - | 2 | 277 | 3488763 | ||
BP753919.1 | - | 2 | 576 | |||
DQ372939.1 | - | 2 | 575 |