Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027436.3 | NP_081712.2 | 13 | 1561 | ||
GenBank mRNA | AK017491.1 | - | 13 | 1537 | ||
AK038479.1 | - | 13 | 1555 | |||
AK044460.1 | - | 13 | 1546 | |||
AK076613.1 | - | 13 | 1491 | |||
AK146428.1 | - | 14 | 1552 | |||
AK183152.1 | - | 2 | 139 | |||
AK218084.1 | - | 2 | 193 | |||
AK219345.1 | - | 1 | 101 | |||
BC049871.1 | - | 4 | 401 | |||
BC110358.1 | - | 13 | 1539 | 5322591 | ||
BC144798.1 | - | 13 | 1542 | |||
BC145711.1 | - | 13 | 1542 | 40131014 |