Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027436.3 | NP_081712.2 | 10, 11 | 1220 | ||
GenBank mRNA | AK017491.1 | - | 10, 11 | 1196 | ||
AK038479.1 | - | 10, 11 | 1214 | |||
AK044460.1 | - | 10, 11 | 1205 | |||
AK076613.1 | - | 10, 11 | 1150 | |||
AK146428.1 | - | 11, 12 | 1211 | |||
BC049871.1 | - | 1, 2 | 60 | |||
BC110358.1 | - | 10, 11 | 1198 | 5322591 | ||
BC144798.1 | - | 10, 11 | 1201 | |||
BC145711.1 | - | 10, 11 | 1201 | 40131014 |