Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016155.5 | NP_057239.4 | 9 | 1337 | ||
XM_011538355.2 | XP_011536657.1 | 9 | 1191 | |||
XM_011538356.2 | XP_011536658.1 | 9 | 1187 | |||
XM_011538357.2 | XP_011536659.1 | 9 | 1136 | |||
GenBank mRNA | AB021225.2 | - | 9 | 1343 | ||
BC040507.1 | - | 9 | 1329 | 5301089 | ||
BC045610.1 | - | 9 | 1346 | 5273795 | ||
X89576.1 | - | 8 | 1215 |