Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027438.3 | NP_081714.2 | 1 | 1143 | ||
GenBank mRNA | AK017476.1 | - | 1 | 937 | ||
AK028331.1 | - | 1 | 1081 | |||
AK136933.1 | - | 1 | 1145 | |||
BC116687.1 | - | 1 | 897 | 40106117 | ||
BC116688.1 | - | 1 | 893 | 40106116 |