Search Thermo Fisher Scientific
Gene Symbol: Sik3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027498.3 | NP_081774.3 | 20, 21 | 2811 | ||
GenBank mRNA | AK129257.1 | - | 14, 15 | 1824 | ||
BC058975.1 | - | 3, 4 | 524 | |||
BC063268.1 | - | 20, 21 | 2607 | 4187769 | ||
BC080688.1 | - | 20, 21 | 2753 | 6515742 | ||
BC082313.1 | - | 6, 7 | 966 |