Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177995.1 | NP_001171466.1 | 9 | 1962 | ||
NM_001291026.1 | NP_001277955.1 | 9 | 1956 | |||
NM_001291029.1 | NP_001277958.1 | 9 | 1962 | |||
NM_027504.3 | NP_081780.3 | 9 | 1959 | |||
GenBank mRNA | AB078338.1 | - | 9 | 1851 | ||
AK083829.1 | - | 9 | 1846 | |||
AK173230.1 | - | 1 | 138 | |||
BC053082.1 | - | 9 | 1897 | |||
BC059838.1 | - | 9 | 1940 | 6409778 |