Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vcpip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030104.1 | - | 1 | 2562 | ||
AK122563.1 | - | 1 | 2575 | |||
BC052908.1 | - | 1 | 752 | |||
BC059209.1 | - | 1 | 2581 | 6406239 |