Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gulp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028450.3 | NP_082726.2 | 6 | 536 | ||
GenBank mRNA | AK014093.1 | - | 5 | 285 | ||
AK017798.1 | - | 6 | 514 | |||
AK028490.1 | - | 5 | 452 | |||
AK036670.1 | - | 6 | 536 | |||
BC032154.1 | - | 6 | 707 | 5068339 | ||
BY713087.1 | - | 5 | 283 | |||
DQ786776.1 | - | 5 | 290 |