Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133911.1 | NP_598672.1 | 7 | 1041 | ||
GenBank mRNA | AK014455.1 | - | 7 | 1013 | ||
AK028929.1 | - | 7 | 980 | |||
BC020317.2 | - | 2 | 174 | |||
BC052391.1 | - | 7 | 1041 |
Set Membership: |
![]() |