Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133911.1 | NP_598672.1 | 3 | 538 | ||
GenBank mRNA | AK014455.1 | - | 3 | 510 | ||
AK028929.1 | - | 3 | 477 | |||
BC052391.1 | - | 3 | 538 |
Set Membership: |
![]() |