Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Medag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027519.3 | NP_081795.1 | 5 | 1004 | ||
GenBank mRNA | AB055406.1 | - | 5 | 1036 | ||
AK018128.1 | - | 5 | 820 | |||
AK036605.1 | - | 5 | 1005 | |||
AK042557.1 | - | 5 | 717 | |||
AK047657.1 | - | 5 | 1061 | |||
AK086092.1 | - | 5 | 1013 | |||
AK138268.1 | - | 5 | 703 | |||
AK160904.1 | - | 5 | 1011 | |||
AK161526.1 | - | 5 | 1013 | |||
AK162725.1 | - | 5 | 1005 | |||
AK163617.1 | - | 5 | 703 | |||
AK165529.1 | - | 5 | 801 | |||
BC026658.1 | - | 5 | 767 | |||
BC116246.1 | - | 5 | 926 | 40093530 | ||
BC116247.1 | - | 5 | 924 | 40093536 |