Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Medag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027519.3 | NP_081795.1 | 4 | 841 | ||
GenBank mRNA | AB055406.1 | - | 4 | 873 | ||
AK018128.1 | - | 4 | 657 | |||
AK036605.1 | - | 4 | 842 | |||
AK042557.1 | - | 4 | 554 | |||
AK047657.1 | - | 4 | 898 | |||
AK086092.1 | - | 4 | 850 | |||
AK138268.1 | - | 4 | 540 | |||
AK160904.1 | - | 4 | 848 | |||
AK161526.1 | - | 4 | 850 | |||
AK162725.1 | - | 4 | 842 | |||
AK163617.1 | - | 4 | 540 | |||
AK165529.1 | - | 4 | 638 | |||
BC026658.1 | - | 4 | 604 | |||
BC116246.1 | - | 4 | 763 | 40093530 | ||
BC116247.1 | - | 4 | 761 | 40093536 |