Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027543.3 | NP_081819.2 | 3 | 924 | ||
GenBank mRNA | AY255602.1 | - | 1 | 510 | ||
BC043021.1 | - | 1 | 692 | 5686241 | ||
BC096520.1 | - | 3 | 924 | 5720034 |
Set Membership: |
![]() |