Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027551.2 | NP_081827.1 | 5 | 1162 | ||
GenBank mRNA | AK014532.1 | - | 1 | 29 | ||
AK086025.1 | - | 5 | 1163 | |||
AK146580.1 | - | 5 | 1219 | |||
AK148398.1 | - | 5 | 1171 | |||
BC115428.1 | - | 4 | 1105 | 40094085 | ||
BC115429.1 | - | 4 | 1105 | 40094088 |