Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080510.2 | NP_536771.2 | 7 | 1458 | ||
GenBank mRNA | AF334958.1 | - | 7 | 1458 | ||
AK014923.1 | - | 7 | 1450 | |||
BC050815.1 | - | 7 | 1450 | 6774293 |