Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027629.3 | NP_081905.1 | 4 | 774 | ||
GenBank mRNA | AK016437.1 | - | 2 | 133 | ||
AK039189.1 | - | 4 | 758 | |||
AK040513.1 | - | 4 | 694 | |||
BC020351.1 | - | 4 | 987 | |||
BC076571.1 | - | 4 | 671 | 30356163 |