Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027630.1 | NP_081906.1 | 5, 6 | 1239 | ||
GenBank mRNA | AK016452.1 | - | 5, 6 | 1240 |