Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glt6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039095.1 | NP_001034184.1 | 5 | 835 | ||
GenBank mRNA | AK016765.1 | - | 5 | 842 | ||
BC111823.1 | - | 5 | 835 | 40052059 |