Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027677.2 | NP_081953.2 | 1 | 1077 | ||
GenBank mRNA | BC085285.1 | - | 1 | 1090 | 30745457 | |
EU076431.1 | - | 1 | 736 |
Set Membership: |
![]() |