Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ACADS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000017.3 | NP_000008.1 | 8 | 1154 | ||
NM_001302554.1 | NP_001289483.1 | 8 | 1142 | |||
GenBank mRNA | AB527081.1 | - | 8 | 1006 | ||
AK300645.1 | - | 8 | 1020 | |||
BC025963.1 | - | 8 | 1053 | 4842286 | ||
M26393.1 | - | 8 | 1038 |