Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Otud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027715.1 | NP_081991.1 | 1 | 1277 | ||
GenBank mRNA | AK016972.1 | - | 1 | 584 | ||
AK206709.1 | - | 1 | 231 |