Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175168.4 | NP_780377.1 | 8, 9 | 1529 | ||
GenBank mRNA | AF531873.1 | - | 8, 9 | 1379 | ||
AK053044.1 | - | 8, 9 | 1462 | |||
AK163135.1 | - | 8, 9 | 1462 | |||
BC076578.1 | - | 8, 9 | 1529 | 30619280 |