Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ubiad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027873.2 | NP_082149.1 | 1 | 418 | ||
GenBank mRNA | AK004550.1 | - | 1 | 418 | ||
AK034787.1 | - | 1 | 445 | |||
AK078442.2 | - | 1 | 419 | |||
BC015303.1 | - | 1 | 126 | 4241204 |