- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Cdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027879.2 | NP_082155.1 | 3 | 462 | ||
GenBank mRNA | AK004569.1 | - | 3 | 429 | ||
AK040680.1 | - | 3 | 412 | |||
AK084140.1 | - | 3 | 460 | |||
AK135411.1 | - | 3 | 429 | |||
BC061220.1 | - | 3 | 437 | |||
BC147503.1 | - | 3 | 462 | 9056386 | ||
BC158078.1 | - | 3 | 462 | 9088170 |