Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Telo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163661.1 | NP_001157133.1 | 18 | 2718 | ||
NM_027880.2 | NP_082156.2 | 18 | 2629 | |||
GenBank mRNA | AK004582.1 | - | 18 | 2499 | ||
AK028658.1 | - | 18 | 2483 | |||
AK045321.1 | - | 9 | 3542 | |||
AK050773.1 | - | 18 | 2448 | |||
AK129189.1 | - | 16 | 3317 | |||
AK143827.1 | - | 18 | 2719 | |||
AK146963.1 | - | 18 | 2505 | |||
AK154727.1 | - | 18 | 2531 | |||
BC011077.1 | - | 18 | 2479 | 3984911 |