Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163645.1 | NP_001157117.1 | 17 | 2367 | ||
NM_027881.3 | NP_082157.2 | 17 | 2085 | |||
GenBank mRNA | AK004768.1 | - | 17 | 2087 | ||
AK037114.1 | - | 17 | 2054 | |||
AK173006.1 | - | 17 | 5235 | |||
BC044866.1 | - | 16 | 2012 | 5003631 | ||
BC086485.1 | - | 17 | 2279 | 5344992 |