Gene Symbol: Trnau1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027925.3 | NP_082201.2 | 5 | 508 | ||
GenBank mRNA | AK003534.1 | - | 4 | 1171 | ||
AK152986.1 | - | 5 | 457 | |||
AK215622.1 | - | 1 | 32 | |||
BC005795.1 | - | 5 | 465 | |||
BC048840.1 | - | 5 | 459 | 5008141 | ||
BC055454.1 | - | 5 | 471 | 4014366 | ||
BC099594.1 | - | 3 | 193 |