Gene Symbol: Trnau1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027925.3 | NP_082201.2 | 7 | 727 | ||
GenBank mRNA | AK003534.1 | - | 6 | 1390 | ||
AK152986.1 | - | 7 | 676 | |||
BC005795.1 | - | 7 | 684 | |||
BC048840.1 | - | 7 | 678 | 5008141 | ||
BC055454.1 | - | 7 | 690 | 4014366 | ||
BC099594.1 | - | 5 | 412 |