- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Trnau1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027925.3 | NP_082201.2 | 5, 6 | 578 | ||
GenBank mRNA | AK003534.1 | - | 4, 5 | 1241 | ||
AK152986.1 | - | 5, 6 | 527 | |||
AK215622.1 | - | 1, 2 | 102 | |||
BC005795.1 | - | 5, 6 | 535 | |||
BC048840.1 | - | 5, 6 | 529 | 5008141 | ||
BC055454.1 | - | 5, 6 | 541 | 4014366 | ||
BC099594.1 | - | 3, 4 | 263 |