Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027959.3 | NP_082235.1 | 4 | 352 | ||
GenBank mRNA | AK005989.1 | - | 4 | 358 | ||
AK076558.1 | - | 4 | 353 | |||
AK133792.1 | - | 4 | 359 | |||
AK151008.1 | - | 4 | 333 | |||
AK152960.1 | - | 4 | 365 | |||
AK165877.1 | - | 4 | 331 | |||
AK166111.1 | - | 4 | 359 | |||
AK167386.1 | - | 4 | 370 | |||
AK168136.1 | - | 4 | 359 | |||
AK168279.1 | - | 4 | 359 | |||
AK192703.1 | - | 3 | 261 | |||
BC006865.1 | - | 4 | 341 | 2645183 |