Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162883.1 | NP_001156355.1 | 3 | 301 | ||
NM_173786.1 | NP_776147.1 | 3 | 353 | |||
GenBank mRNA | AI530062.1 | - | 3 | 353 | ||
BC020489.1 | - | 3 | 331 | 2649139 | ||
BC057684.1 | - | 3 | 361 | 3675306 | ||
CJ058381.1 | - | 3 | 302 |
Gene Symbol: Apol9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168660.1 | NP_001162131.1 | 3 | 372 | ||
NM_173743.4 | NP_776104.1 | 3 | 323 | |||
GenBank mRNA | AK009386.1 | - | 3 | 373 | ||
AK050167.1 | - | 3 | 325 |