Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024246.5 | NP_077208.1 | 2 | 375 | ||
GenBank mRNA | AK009760.1 | - | 2 | 376 | ||
AK010144.1 | - | 2 | 467 | |||
BC003309.1 | - | 2 | 430 | 3154669 |