Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101433.1 | NP_001094903.1 | 3 | 595 | ||
GenBank mRNA | BC034735.1 | - | 1 | 149 | ||
BC147263.1 | - | 3 | 685 | 9056350 | ||
BC147264.1 | - | 3 | 685 | 9056314 | ||
BU701574.1 | - | 1 | 83 | |||
CN526234.1 | - | 3 | 375 |