Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028032.3 | NP_082308.1 | 10 | 1508 | ||
GenBank mRNA | AK090027.1 | - | 10 | 1494 | ||
AK161507.1 | - | 10 | 1466 | |||
AK161704.1 | - | 10 | 1457 | |||
AK164497.1 | - | 10 | 1492 | |||
AK167189.1 | - | 10 | 1497 | |||
AK167530.1 | - | 10 | 1456 | |||
AK220192.1 | - | 10 | 1461 | |||
BC022916.1 | - | 11 | 1281 | |||
BC038824.1 | - | 4 | 502 | |||
BC065100.1 | - | 10 | 1481 | 30535680 | ||
BC080692.1 | - | 10 | 1431 | 30431716 |