Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028049.2 | NP_082325.2 | 6 | 844 | ||
GenBank mRNA | AK002745.1 | - | 6 | 724 | ||
AK005473.1 | - | 5 | 672 | |||
AK179843.1 | - | 1 | 45 | |||
AK181887.1 | - | 1 | 65 | |||
AK182889.1 | - | 2 | 279 | |||
AK187855.1 | - | 1 | 48 | |||
AK188724.1 | - | 2 | 279 | |||
AK188900.1 | - | 1 | 48 | |||
AK189126.1 | - | 2 | 182 | |||
AK207864.1 | - | 2 | 282 | |||
AK211102.1 | - | 2 | 146 | |||
AK213133.1 | - | 2 | 279 | |||
AK219248.1 | - | 2 | 279 | |||
BC018273.1 | - | 6 | 784 | 4458296 |