Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF361644.1 | - | 6 | 760 | ||
AK005532.1 | - | 6 | 743 | |||
AK010702.1 | - | 4 | 521 | |||
AK031742.1 | - | 6 | 598 | |||
AK049358.1 | - | 6 | 746 | |||
AK082340.1 | - | 4 | 526 | |||
AK082749.1 | - | 4 | 362 | |||
AK085617.1 | - | 6 | 753 | |||
AK086940.1 | - | 6 | 693 | |||
AK143428.1 | - | 6 | 736 | |||
AK215627.1 | - | 2 | 82 | |||
AK220209.1 | - | 6 | 570 | |||
BC013549.1 | - | 6 | 762 | 4222133 |