Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111025.1 | NP_001104495.1 | 6 | 819 | ||
NM_024464.3 | NP_077784.2 | 6 | 825 | |||
GenBank mRNA | AK008583.2 | - | 6 | 827 | ||
BC002202.1 | - | 4 | 469 | 3489499 |